Merck
  • Mesothelial mobilization in the developing lung and heart differs in timing, quantity, and pathway dependency.
Mesothelial mobilization in the developing lung and heart differs in timing, quantity, and pathway dependency.
American journal of physiology. Lung cellular and molecular physiology (2019-02-01)
Timo H Lüdtke, Carsten Rudat, Jennifer Kurz, Regine Häfner, Franziska Greulich, Irina Wojahn, Nurullah Aydoğdu, Tamrat M Mamo, Marc-Jens Kleppa, Mark-Oliver Trowe, Tobias Bohnenpoll, Makoto Mark Taketo, Andreas Kispert
ABSTRACT
The mesothelial lining of the lung, the visceral pleura, and of the heart, the epicardium, derive from a common multipotent precursor tissue, the mesothelium of the embryonic thoracic cavity that also contributes to organ-specific mesenchymal cell types. Insight into mesothelial mobilization and differentiation has prevailedin the developing heart while the mesenchymal transition and fate of the visceral pleura are poorly understood. Here, we use the fact that the early mesothelium of both the lung and the heart expresses the transcription factor gene Wt1, to comparatively analyze mesothelial mobilization in the two organs by a genetic cre-loxP-based conditional approach. We show that epicardial cells are mobilized in a large number between E12.5 and E14.5, whereas pleural mobilization occurs only sporadically and variably in few regions of the lung in a temporally highly confined manner shortly after E12.5. Mesothelium-specific inactivation of unique pathway components using a Wt1creERT2 line excluded a requirement for canonical WNT, NOTCH, HH, TGFB, PDGFRA, and FGFR1/FGFR2 signaling in the mesenchymal transition of the visceral pleura but indicated a deleterious effect of activated WNT, NOTCH, and HH signaling on lung development. Epicardial mobilization was negatively impacted on by loss of HH, PDGFRA, FGFR1/2 signaling. Epicardial overactivation of WNT, NOTCH, and HH disturbed epicardial and myocardial integrity. We conclude that mesothelial mobilization in the developing lung and heart differs in timing, quantity and pathway dependency, indicating the organ specificity of the program.
Product Description
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)