Merck
  • High possibility of hepatocarcinogenesis in HBV genotype C1 infected Cambodians is indicated by 340 HBV C1 full-genomes analysis from GenBank.
High possibility of hepatocarcinogenesis in HBV genotype C1 infected Cambodians is indicated by 340 HBV C1 full-genomes analysis from GenBank.
Scientific reports (2019-08-23)
Channarena Chuon, Kazuaki Takahashi, Junko Matsuo, Keiko Katayama, Chikako Yamamoto, Ko Ko, Sirany Hok, Shintaro Nagashima, Sheikh Mohammad Fazle Akbar, Junko Tanaka
ABSTRACT
Approximately 75% of hepatocellular carcinomas (HCC) occur in Asia; core promoter mutations are associated with HCC in HBV genotype C, the dominant genotype in Cambodia. We analyzed these mutations in Cambodian residents and compared them with HBV full genomes registered in GenBank. We investigated the characteristics of 26 full-length HBV genomes among 35 residents positive for hepatitis B surface antigen in Siem Reap province, Cambodia. Genotype C1 was dominant (92.3%, 24/26), with one case of B2 and B4 each. Multiple mutations were confirmed in 24 Cambodian C1 isolates, especially double mutation at A1762T/G1764A in 18 isolates (75.0%), and combination mutation at C1653T and/or T1753V and A1762T/G1764A in 14 isolates (58.3%). In phylogenetic analysis, 16 of 24 isolates were located in the cluster with Laos, Thailand, and Malaysia. In 340 GenBank-registered C1 strains, 113 (33.2%) had combination mutation amongst which 16.5%, 34.2%, and 95.2% were found in ASC, chronic hepatitis, and liver cirrhosis (LC)/HCC respectively (P < 0. 001). Mutations were abundantly found in 24 Cambodian C1 isolates, and 340 C1 strains from GenBank showed mutation in genotype C1 brings high possibility of LC/HCC occurrence. Therefore, we suggest that Cambodian people infected with HBV genotype C1 have high possibility of hepatocarcinogenesis.