Merck
  • Structures of Ebola and Reston Virus VP35 Oligomerization Domains and Comparative Biophysical Characterization in All Ebolavirus Species.
Structures of Ebola and Reston Virus VP35 Oligomerization Domains and Comparative Biophysical Characterization in All Ebolavirus Species.
Structure (London, England : 1993) (2018-11-30)
Luca Zinzula, István Nagy, Massimiliano Orsini, Elisabeth Weyher-Stingl, Andreas Bracher, Wolfgang Baumeister
ABSTRACT
The multifunctional virion protein 35 (VP35) of ebolaviruses is a critical determinant of virulence and pathogenesis indispensable for viral replication and host innate immune evasion. Essential for VP35 function is homo-oligomerization via a coiled-coil motif. Here we report crystal structures of VP35 oligomerization domains from the prototypic Ebola virus (EBOV) and the non-pathogenic Reston virus (RESTV), together with a comparative biophysical characterization of the domains from all known species of the Ebolavirus genus. EBOV and RESTV VP35 oligomerization domains form bipartite parallel helix bundles with a canonical coiled coil in the N-terminal half and increased plasticity in the highly conserved C-terminal half. The domain assembles into trimers and tetramers in EBOV, whereas it exclusively forms tetramers in all other ebolavirus species. Substitution of coiled-coil leucine residues critical for immune antagonism leads to aberrant oligomerization. A conserved arginine involved in inter-chain salt bridges stabilizes the VP35 oligomerization domain and modulates between coiled-coil oligomeric states.