Merck
  • The X-Linked DDX3X RNA Helicase Dictates Translation Reprogramming and Metastasis in Melanoma.
The X-Linked DDX3X RNA Helicase Dictates Translation Reprogramming and Metastasis in Melanoma.
Cell reports (2019-06-20)
Bengt Phung, Maciej Cieśla, Adriana Sanna, Nicola Guzzi, Giulia Beneventi, Phuong Cao Thi Ngoc, Martin Lauss, Rita Cabrita, Eugenia Cordero, Ana Bosch, Frida Rosengren, Jari Häkkinen, Klaus Griewank, Annette Paschen, Katja Harbst, Håkan Olsson, Christian Ingvar, Ana Carneiro, Hensin Tsao, Dirk Schadendorf, Kristian Pietras, Cristian Bellodi, Göran Jönsson
ABSTRACT
The X-linked DDX3X gene encodes an ATP-dependent DEAD-box RNA helicase frequently altered in various human cancers, including melanomas. Despite its important roles in translation and splicing, how DDX3X dysfunction specifically rewires gene expression in melanoma remains completely unknown. Here, we uncover a DDX3X-driven post-transcriptional program that dictates melanoma phenotype and poor disease prognosis. Through an unbiased analysis of translating ribosomes, we identified the microphthalmia-associated transcription factor, MITF, as a key DDX3X translational target that directs a proliferative-to-metastatic phenotypic switch in melanoma cells. Mechanistically, DDX3X controls MITF mRNA translation via an internal ribosome entry site (IRES) embedded within the 5' UTR. Through this exquisite translation-based regulatory mechanism, DDX3X steers MITF protein levels dictating melanoma metastatic potential in vivo and response to targeted therapy. Together, these findings unravel a post-transcriptional layer of gene regulation that may provide a unique therapeutic vulnerability in aggressive male melanomas.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-MLANA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)