Merck
  • Freezing and melting of lipid bilayers and the mode of action of nonactin, valinomycin, and gramicidin.
Freezing and melting of lipid bilayers and the mode of action of nonactin, valinomycin, and gramicidin.
Science (New York, N.Y.) (1971-10-22)
S Krasne, G Eisenman, G Szabo
ABSTRACT
An abrupt loss of effectiveness of the presumed carriers, nonactin and valinomycin, in mediating ion conductance occurred at the same temperature as the membrane fluidity, judged visually, was lost. By contrast, the effects of the presumed channel-former, gramicidin, were the same on solid and liquid membranes. Taken together, these findings imply that freezing the membrane primarily reduces the mobility of these antibiotics with little effect on their solubility.
Sigma-Aldrich
Nonactin, from Streptomyces sp., ≥97.0% (TLC)
Sigma-Aldrich
Nonactin, from Streptomyces griseus, ≥98.0% (Total homologs, HPLC)