Merck
Specific binding of arginine to TAR RNA.
Proceedings of the National Academy of Sciences of the United States of America (1992-04-01)
J Tao, A D Frankel
ABSTRACT
A single arginine residue within the basic region of the human immunodeficiency virus Tat protein mediates specific binding of Tat peptides to a three-nucleotide bulge in TAR RNA. It has been proposed that arginine recognizes TAR by forming a network of hydrogen bonds with two structurally distinct phosphates, an interaction termed the "arginine fork." Here it is shown that L-arginine blocks the Tat peptide/TAR interaction, whereas L-lysine and analogs of arginine that remove specific hydrogen bond donors do not. Experiments using an L-arginine affinity column demonstrate that arginine and the Tat peptides bind to the same site in TAR. Modification of two phosphates located at the junction of the double-stranded stem and bulge and modification of two adenine N7 groups in base-paired regions of TAR interfere with specific arginine binding. The results emphasize the importance of RNA structure in RNA-protein recognition and provide methods to identify arginine-binding sites in RNAs.
Sigma-Aldrich
L-Lysine–Agarose, lyophilized powder