Merck
  • Raffinose metabolism in Escherichia coli K12. Purification and properties of a new alpha-galactosidase specified by a transmissible plasmid.
Raffinose metabolism in Escherichia coli K12. Purification and properties of a new alpha-galactosidase specified by a transmissible plasmid.
European journal of biochemistry (1976-08-01)
K Schmid, R Schmitt
PMID786627
ABSTRACT
The utilization by Escherichia coli K12 of raffinose as sole carbon source depends on a new raffinose transport system, an invertase and an alpha-galactosidase specified by the Raf-plasmid D1021. The alpha-galactosidase was purified to homogeneity from a mutant strain with constitutive synthesis of the enzyme. alpha-Galactosidase hydrolyzes p-nitrophenyl-alpha-D-galactoside (Km 0.14 mM), methyl-alpha-D-galactoside (Km 30mM), melibiose (Km 3.2 mM) and raffinose (Km 60 mM). The enzymatic activity is strongly inhibited by Ag+, p-chloromercuriphenyl sulfonic acid and, to a lesser extent, by iodoacetamide. Isoelectric focusing indicates the existence of one form of alpha-galactosidase with an isoelectric point of 5.1. The purified enzyme has an sw,20 value of 11.7 +/- 0.3S and a molecular weight of 329000 +/- 4000; this value is not reduced at high dilutions. When examined by dodecylsulphate gel electrophoresis, purified alpha-galactosidase yields a single subunit band of molecular weight 82000 suggesting that the intact enzyme consists of four subunits. Amino acid analysis indicates the presence of approximately 712 amino acid residues per quarter molecule including 8 half-cystine residues. No carbohydrate moiety has been detected. High resolution electron micrographs and Markham rotation of alpha-galactosidase show enzyme molecules of approximately 11 x 11 nm containing four globular subunits in a tetragonal arrangement. The plasmid-coded alpha-galactosidase differs from the homologous E. coli enzyme by substrate affinities, cofactor requirements, stability and toluene resistance. It can, therefore, be used as a marker enzyme suitable for the detection in vivo of Raf-plasmids.
Sigma-Aldrich
α-Galactosidase, positionally specific from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution
Pricing and availability is not currently available.