Merck
  • Automated kinetic determination of angiotensin-converting enzyme in serum.
Automated kinetic determination of angiotensin-converting enzyme in serum.
Clinical chemistry (1984-06-01)
A Harjanne
ABSTRACT
In this automated kinetic modification of a previous method (Anal Biochem 95: 540-548, 1979) for determining angiotensin-converting enzyme (EC 3.4.15.1), 3-(2- furylacryloyl )-L- phenylalanylglycylglycine is used as the substrate. The change in absorbance at 340 nm is used to monitor hydrolysis of the substrate. The rate of hydrolysis is roughly threefold greater than with previously reported substrates, so assay time and sensitivity are improved.
Sigma-Aldrich
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly
Pricing and availability is not currently available.