Merck
  • Synthesis and biological evaluation of 3-arylcoumarin derivatives as potential anti-diabetic agents.
Synthesis and biological evaluation of 3-arylcoumarin derivatives as potential anti-diabetic agents.
Journal of enzyme inhibition and medicinal chemistry (2018-10-27)
Yuheng Hu, Bing Wang, Jie Yang, Teng Liu, Jie Sun, Xiaojing Wang
ABSTRACT
A variety of substituted 3-arylcoumarin derivatives were synthesised through microwave radiation heating. The method has characteristics of environmental friendliness, economy, simple separation, and purification process, less by-products and high reaction yield. Those 3-arylcoumarin derivatives were screened for antioxidant, α-glucosidase inhibitory and advanced glycation end-products (AGEs) formation inhibitory. Most compounds exhibited significant antioxidant and AGEs formation inhibitory activities. Anti-diabetic activity studies showed that compounds 11 and 17 were equipotent to the standard drug glibenclamide in vivo. According to the experimental results, the target compound 35 can be used as a lead compound for the development of new anti-diabetic drugs. The whole experiment showed that anti-diabetic activity is prevalent in 3-arylcoumarins, which added a new natural skeleton to the development of anti-diabetic active drugs.
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein
Pricing and availability is not currently available.