Merck
Lipidomic Analysis.
Analytical chemistry (2018-03-16)
Michal Holčapek, Gerhard Liebisch, Kim Ekroos
ABSTRACT
The state-of-art in the lipidomic analysis is summarized here to provide the overview of available sample preparation strategies, mass spectrometry (MS)-based methods for the qualitative analysis of lipids, and the quantitative MS approaches for high-throughput clinical workflows. Major challenges in terms of widely accepted best practices for lipidomic analysis, nomenclature, and standards for data reporting are discussed as well.
Product Description
Avanti
SPLASH LIPIDOMIX Mass Spec Standard, Avanti Research - A Croda Brand