Merck
  • Preparation and reconstitution with divalent metal ions of class I and class II Clostridium histolyticum apocollagenases.
Preparation and reconstitution with divalent metal ions of class I and class II Clostridium histolyticum apocollagenases.
Biochemistry (1988-09-20)
E L Angleton, H E Van Wart
ABSTRACT
Both gamma- and zeta-collagenases from Clostridium histolyticum are fully and reversibly inhibited by 1,10-phenanthroline at pH 7.5 in the presence of 10 mM CaCl2 with KI values of 0.11 and 0.040 mM, respectively. The inhibition is caused by removal of the single, active-site Zn(II) present in each of these enzymes. The nonchelating analogue 1,5-phenanthroline has no effect on the activity of either enzyme. Dialysis of the enzymes in the presence of 1,10-phenanthroline, followed by back dialysis against buffer containing no chelating agent, gives the respective apocollagenases. Both apoenzymes can be instantaneously and fully reactivated by the addition of 1 equiv of Zn(II). Variable amounts of activity are restored to both apocollagenases by Co(II) and Ni(II) and to gamma-apocollagenase by Cu(II). The activity titration curve for gamma-apocollagenase with Co(II) and Scatchard plots for the reconstitution of gamma-apocollagenase with Cu(II) and Ni(II) and of zeta-apocollagenase with Ni(II) and Co(II) indicate that all activity changes are the result of binding of a single equivalent of these divalent metal ions at the active site of the collagenases. Cd(II) and Hg(II) do not restore measurable activity to either apoenzyme.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid