Merck
  • Up-regulated miR-29c inhibits cell proliferation and glycolysis by inhibiting SLC2A3 expression in prostate cancer.
Up-regulated miR-29c inhibits cell proliferation and glycolysis by inhibiting SLC2A3 expression in prostate cancer.
Gene (2018-05-02)
Junliang Li, Fangqiu Fu, Xuechao Wan, Shengsong Huang, Denglong Wu, Yao Li
ABSTRACT
Prostate cancer (PCa) is the most commonly cancer in male worldwide. However, the molecular mechanisms underlying the progression of PCa remain unclear. MiR-29c was reported to be down-regulated in several kinds of tumors. Here, we for the first time demonstrated miR-29c was down-regulated in PCa samples. SLC2A3, a regulator of glycolysis, was validated as a direct target of miR-29c. Moreover, functional studies showed miR-29c could inhibit cell growth, induce apoptosis and deceased the rate of glucose metabolism. Accordingly, we identified miR-29c acted as a tumor-suppressor and was down-regulated in PCa. We thought this study will provide useful information to explore the potential candidate biomarkers for diagnosis and prognosis targets of PCa.
Sigma-Aldrich
Lactate Assay Kit, sufficient for 100 colorimetric or fluorometric tests