Merck
  • The major β-catenin/E-cadherin junctional binding site is a primary molecular mechano-transductor of differentiation in vivo.
The major β-catenin/E-cadherin junctional binding site is a primary molecular mechano-transductor of differentiation in vivo.
eLife (2018-07-20)
Jens-Christian Röper, Démosthène Mitrossilis, Guillaume Stirnemann, François Waharte, Isabel Brito, Maria-Elena Fernandez-Sanchez, Marc Baaden, Jean Salamero, Emmanuel Farge
ABSTRACT
In vivo, the primary molecular mechanotransductive events mechanically initiating cell differentiation remain unknown. Here we find the molecular stretching of the highly conserved Y654-β-catenin-D665-E-cadherin binding site as mechanically induced by tissue strain. It triggers the increase of accessibility of the Y654 site, target of the Src42A kinase phosphorylation leading to irreversible unbinding. Molecular dynamics simulations of the β-catenin/E-cadherin complex under a force mimicking a 6 pN physiological mechanical strain predict a local 45% stretching between the two α-helices linked by the site and a 15% increase in accessibility of the phosphorylation site. Both are quantitatively observed using FRET lifetime imaging and non-phospho Y654 specific antibody labelling, in response to the mechanical strains developed by endogenous and magnetically mimicked early mesoderm invagination of gastrulating Drosophila embryos. This is followed by the predicted release of 16% of β-catenin from junctions, observed in FRAP, which initiates the mechanical activation of the β-catenin pathway process.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human GALC