Merck
  • Expeditious synthesis and cytotoxic activity of new cyanoindolo[3,2-c]quinolines and benzimidazo[1,2-c]quinazolines.
Expeditious synthesis and cytotoxic activity of new cyanoindolo[3,2-c]quinolines and benzimidazo[1,2-c]quinazolines.
Bioorganic & medicinal chemistry letters (2000-09-30)
C Lamazzi, S Léonce, B Pfeiffer, P Renard, G Guillaumet, C W Rees, T Besson
ABSTRACT
Novel 6-cyanoindolo[3,2-c]quinoline and 6-cyanobenzimidazo[1,2-c]quinazoline derivatives have been synthesised by treatment of the appropriate aromatic amines with 4.5-dichloro-1,2,3-dithiazolium chloride 1 (Appel salt). The cytotoxicity and the effect of these compounds on cellular growth were measured.