Merck
  • Mitochondria Restrict Growth of the Intracellular Parasite Toxoplasma gondii by Limiting Its Uptake of Fatty Acids.
Mitochondria Restrict Growth of the Intracellular Parasite Toxoplasma gondii by Limiting Its Uptake of Fatty Acids.
Cell metabolism (2018-04-05)
Lena Pernas, Camilla Bean, John C Boothroyd, Luca Scorrano
ABSTRACT
How intracellular pathogens acquire essential non-diffusible host metabolites and whether the host cell counteracts the siphoning of these nutrients by its invaders are open questions. Here we show that host mitochondria fuse during infection by the intracellular parasite Toxoplasma gondii to limit its uptake of fatty acids (FAs). A combination of genetics and imaging of FA trafficking indicates that Toxoplasma infection triggers lipophagy, the autophagy of host lipid droplets (LDs), to secure cellular FAs essential for its proliferation. Indeed, Toxoplasma FA siphoning and growth are reduced in host cells genetically deficient for autophagy or triglyceride depots. Conversely, Toxoplasma FA uptake and proliferation are increased in host cells lacking mitochondrial fusion, required for efficient mitochondrial FA oxidation, or where mitochondrial FA oxidation is pharmacologically inhibited. Thus, mitochondrial fusion can be regarded as a cellular defense mechanism against intracellular parasites, by limiting Toxoplasma access to host nutrients liberated by lipophagy.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Atglistatin, ≥98% (HPLC)
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)