Merck
  • The Protease Activated Receptor2 Promotes Rab5a Mediated Generation of Pro-metastatic Microvesicles.
The Protease Activated Receptor2 Promotes Rab5a Mediated Generation of Pro-metastatic Microvesicles.
Scientific reports (2018-05-11)
Kaushik Das, Ramesh Prasad, Sreetama Roy, Ashis Mukherjee, Prosenjit Sen
ABSTRACT
Metastasis, the hallmark of cancer propagation is attributed by the modification of phenotypic/functional behavior of cells to break attachment and migrate to distant body parts. Cancer cell-secreted microvesicles (MVs) contribute immensely in disease propagation. These nano-vesicles, generated from plasma membrane outward budding are taken up by nearby healthy cells thereby inducing phenotypic alterations in those recipient cells. Protease activated receptor 2 (PAR2), activated by trypsin, also contributes to cancer progression by increasing metastasis, angiogenesis etc. Here, we report that PAR2 activation promotes pro-metastatic MVs generation from human breast cancer cell line, MDA-MB-231. Rab5a, located at the plasma membrane plays vital roles in MVs biogenesis. We show that PAR2 stimulation promotes AKT phosphorylation which activates Rab5a by converting inactive Rab5a-GDP to active Rab5a-GTP. Active Rab5a polymerizes actin which critically regulates MVs shedding. Not only MVs generation, has this Rab5a activation also promoted cell migration and invasion. We reveal that Rab5a is over-expressed in human breast tumor specimen and contributes MVs generation in those patients. The involvement of p38 MAPK in MVs-induced cell metastasis has also been highlighted in the present study. Blockade of Rab5a activation can be a potential therapeutic approach to restrict MVs shedding and associated breast cancer metastasis.
Sigma-Aldrich
Ser-Leu-Ile-Gly-Lys-Val-amide, ≥95% (HPLC), solid
Pricing and availability is not currently available.