Merck
  • Synthesis and metabotropic glutamate receptor activity of S-oxidized variants of (-)-4-amino-2-thiabicyclo-[3.1.0]hexane-4,6-dicarboxylate: identification of potent, selective, and orally bioavailable agonists for mGlu2/3 receptors.
Synthesis and metabotropic glutamate receptor activity of S-oxidized variants of (-)-4-amino-2-thiabicyclo-[3.1.0]hexane-4,6-dicarboxylate: identification of potent, selective, and orally bioavailable agonists for mGlu2/3 receptors.
Journal of medicinal chemistry (2007-01-19)
James A Monn, Steven M Massey, Matthew J Valli, Steven S Henry, Gregory A Stephenson, Mark Bures, Marc Hérin, John Catlow, Deborah Giera, Rebecca A Wright, Bryan G Johnson, Sherri L Andis, Ann Kingston, Darryle D Schoepp
ABSTRACT
(-)-4-Amino-2-thiabicyclo-[3.1.0]hexane-4,6-dicarboxylate (LY389795, (-)-3) is a highly potent and selective agonist of metabotropic glutamate receptors 2 (mGlu2) and 3 (mGlu3). As part of our ongoing research program, we have prepared S-oxidized variants of (-)-3, compounds (-)-10, (+)-11 (LY404040), and (-)-12 (LY404039). Each of these chiral heterobicyclic amino acids displaced specific binding of the mGlu2/3 receptor antagonist 3H-2S-2-amino-2-(1S,2S-2-carboxycycloprop-1-yl)-3-(xanth-9-yl)propanoic acid (3H-LY341495) from membranes expressing recombinant human mGlu2 or mGlu3 and acted as potent agonists in cells expressing these receptor subtypes. Docking of the most potent of these derivatives, (+)-11, to mGlu2 revealed the possibility of an additional H-bond interaction between the sulfoxide oxygen of (+)-11 with tyrosine residue Y236. Pharmacokinetic analysis of mGlu active enantiomers (+)-11 and (-)-12 in rats showed each to be well absorbed following oral administration. Consistent with their mGlu2/3 agonist potency and pharmacokinetic properties, both (+)-11 and (-)-12 blocked phencyclidine-evoked ambulations in a dose-dependent manner, indicating their potential as nonclassical antipsychotic agents.
Sigma-Aldrich
LY404039, ≥98% (HPLC)