Merck
  • Identification and Characterization of Von Hippel-Lindau-Recruiting Proteolysis Targeting Chimeras (PROTACs) of TANK-Binding Kinase 1.
Identification and Characterization of Von Hippel-Lindau-Recruiting Proteolysis Targeting Chimeras (PROTACs) of TANK-Binding Kinase 1.
Journal of medicinal chemistry (2017-07-12)
Andrew P Crew, Kanak Raina, Hanqing Dong, Yimin Qian, Jing Wang, Dominico Vigil, Yevgeniy V Serebrenik, Brian D Hamman, Alicia Morgan, Caterina Ferraro, Kam Siu, Taavi K Neklesa, James D Winkler, Kevin G Coleman, Craig M Crews
ABSTRACT
Proteolysis targeting chimeras (PROTACs) are bifunctional molecules that recruit an E3 ligase to a target protein to facilitate ubiquitination and subsequent degradation of that protein. While the field of targeted degraders is still relatively young, the potential for this modality to become a differentiated and therapeutic reality is strong, such that both academic and pharmaceutical institutions are now entering this interesting area of research. In this article, we describe a broadly applicable process for identifying degrader hits based on the serine/threonine kinase TANK-binding kinase 1 (TBK1) and have generalized the key structural elements associated with degradation activities. Compound 3i is a potent hit (TBK1 DC50 = 12 nM, Dmax = 96%) with excellent selectivity against a related kinase IKKε, which was further used as a chemical tool to assess TBK1 as a target in mutant K-Ras cancer cells.
Sigma-Aldrich
(S,S,S)-AHPC hydrochloride, ≥97%
Pricing and availability is not currently available.