Merck
  • Mutations in MAPKBP1 Cause Juvenile or Late-Onset Cilia-Independent Nephronophthisis.
Mutations in MAPKBP1 Cause Juvenile or Late-Onset Cilia-Independent Nephronophthisis.
American journal of human genetics (2017-01-17)
Maxence S Macia, Jan Halbritter, Marion Delous, Cecilie Bredrup, Arthur Gutter, Emilie Filhol, Anne E C Mellgren, Sabine Leh, Albane Bizet, Daniela A Braun, Heon Y Gee, Flora Silbermann, Charline Henry, Pauline Krug, Christine Bole-Feysot, Patrick Nitschké, Dominique Joly, Philippe Nicoud, André Paget, Heidi Haugland, Damien Brackmann, Nayir Ahmet, Richard Sandford, Nurcan Cengiz, Per M Knappskog, Helge Boman, Bolan Linghu, Fan Yang, Edward J Oakeley, Pierre Saint Mézard, Andreas W Sailer, Stefan Johansson, Eyvind Rødahl, Sophie Saunier, Friedhelm Hildebrandt, Alexandre Benmerah
ABSTRACT
Nephronophthisis (NPH), an autosomal-recessive tubulointerstitial nephritis, is the most common cause of hereditary end-stage renal disease in the first three decades of life. Since most NPH gene products (NPHP) function at the primary cilium, NPH is classified as a ciliopathy. We identified mutations in a candidate gene in eight individuals from five families presenting late-onset NPH with massive renal fibrosis. This gene encodes MAPKBP1, a poorly characterized scaffolding protein for JNK signaling. Immunofluorescence analyses showed that MAPKBP1 is not present at the primary cilium and that fibroblasts from affected individuals did not display ciliogenesis defects, indicating that MAPKBP1 may represent a new family of NPHP not involved in cilia-associated functions. Instead, MAPKBP1 is recruited to mitotic spindle poles (MSPs) during the early phases of mitosis where it colocalizes with its paralog WDR62, which plays a key role at MSP. Detected mutations compromise recruitment of MAPKBP1 to the MSP and/or its interaction with JNK2 or WDR62. Additionally, we show increased DNA damage response signaling in fibroblasts from affected individuals and upon knockdown of Mapkbp1 in murine cell lines, a phenotype previously associated with NPH. In conclusion, we identified mutations in MAPKBP1 as a genetic cause of juvenile or late-onset and cilia-independent NPH.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-MAPKBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-V5 antibody produced in rabbit, IgG fraction of antiserum