Merck
  • Transcriptome-based repurposing of apigenin as a potential anti-fibrotic agent targeting hepatic stellate cells.
Transcriptome-based repurposing of apigenin as a potential anti-fibrotic agent targeting hepatic stellate cells.
Scientific reports (2017-03-04)
Daniel F Hicks, Nicolas Goossens, Ana Blas-García, Takuma Tsuchida, Benjamin Wooden, Michael C Wallace, Natalia Nieto, Abigale Lade, Benjamin Redhead, Arthur I Cederbaum, Joel T Dudley, Bryan C Fuchs, Youngmin A Lee, Yujin Hoshida, Scott L Friedman
ABSTRACT
We have used a computational approach to identify anti-fibrotic therapies by querying a transcriptome. A transcriptome signature of activated hepatic stellate cells (HSCs), the primary collagen-secreting cell in liver, and queried against a transcriptomic database that quantifies changes in gene expression in response to 1,309 FDA-approved drugs and bioactives (CMap). The flavonoid apigenin was among 9 top-ranked compounds predicted to have anti-fibrotic activity; indeed, apigenin dose-dependently reduced collagen I in the human HSC line, TWNT-4. To identify proteins mediating apigenin's effect, we next overlapped a 122-gene signature unique to HSCs with a list of 160 genes encoding proteins that are known to interact with apigenin, which identified C1QTNF2, encoding for Complement C1q tumor necrosis factor-related protein 2, a secreted adipocytokine with metabolic effects in liver. To validate its disease relevance, C1QTNF2 expression is reduced during hepatic stellate cell activation in culture and in a mouse model of alcoholic liver injury in vivo, and its expression correlates with better clinical outcomes in patients with hepatitis C cirrhosis (n = 216), suggesting it may have a protective role in cirrhosis progression.These findings reinforce the value of computational approaches to drug discovery for hepatic fibrosis, and identify C1QTNF2 as a potential mediator of apigenin's anti-fibrotic activity.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®