Merck
  • ASP5878, a Novel Inhibitor of FGFR1, 2, 3, and 4, Inhibits the Growth of FGF19-Expressing Hepatocellular Carcinoma.
ASP5878, a Novel Inhibitor of FGFR1, 2, 3, and 4, Inhibits the Growth of FGF19-Expressing Hepatocellular Carcinoma.
Molecular cancer therapeutics (2016-11-12)
Takashi Futami, Hidetsugu Okada, Rumi Kihara, Tatsuya Kawase, Ayako Nakayama, Tomoyuki Suzuki, Minoru Kameda, Nobuaki Shindoh, Tadashi Terasaka, Masaaki Hirano, Sadao Kuromitsu
ABSTRACT
Hepatocellular carcinoma is an aggressive cancer with poor prognosis. Fibroblast growth factor 19, a member of the fibroblast growth factor family, is a ligand for fibroblast growth factor receptor 4. Moreover, it plays a crucial role in the progression of hepatocellular carcinoma. ASP5878 is a novel inhibitor of fibroblast growth factor receptors 1, 2, 3, and 4 that is under development. It inhibits fibroblast growth factor receptor 4 kinase activity with an IC50 of 3.5 nmol/L. ASP5878 potently suppressed the growth of the fibroblast growth factor 19-expressing hepatocellular carcinoma cell lines Hep3B2.1-7, HuH-7, and JHH-7. In the Hep3B2.1-7 cell line, ASP5878 inhibited the phosphorylation of fibroblast growth factor receptor 4 and its downstream signaling molecules as well as induced apoptosis. Oral administration of ASP5878 at 3 mg/kg induced sustained tumor regression in a subcutaneous xenograft mouse model using Hep3B2.1-7. In HuH-7, an orthotopic xenograft mouse model, ASP5878 induced complete tumor regression and dramatically extended the survival of the mice. These results suggest that ASP5878 is a potentially effective therapeutic agent for hepatocellular carcinoma patients with tumors expressing fibroblast growth factor 19. Mol Cancer Ther; 16(1); 68-75. ©2016 AACR.
Sigma-Aldrich
Anti-FGF19 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Pricing and availability is not currently available.