Skip to Content
Merck
Zineb PESTANAL®, analytical standard

Zineb

Zineb PESTANAL®, analytical standard
Synonym(s):
Zinc-ethylenebis(dithiocarbamate)
Empirical Formula (Hill Notation):
C4H6N2S4Zn
CAS Number:
Molecular Weight:
275.75
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing