
Zilpaterol-(isopropyl-13C3) hydrochloride

Synonym(s):
Zilpaterol-13C3 hydrochloride
Empirical Formula (Hill Notation):
13C3C11H19N3O2 · HCl
CAS Number:
Molecular Weight:
300.76
Products
Brand
Formula Weight
Product Line
Format
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing