![Zileuton Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/245/025/8533553c-48df-407c-9682-2d96f377df94/640/8533553c-48df-407c-9682-2d96f377df94.png)
Zileuton Related Compound C
![Zileuton Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/245/025/8533553c-48df-407c-9682-2d96f377df94/640/8533553c-48df-407c-9682-2d96f377df94.png)
Synonym(s):
2-Acetylbenzothiophene, 1-Benzo[b]thiophen-2-yl-ethanone
Empirical Formula (Hill Notation):
C10H8OS
CAS Number:
Molecular Weight:
176.23
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2