Skip to Content
GB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe
Products
Manufacturer
Physical Form
Filter Products
Showing 1-20 of 26
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF83693885
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 15mm disks, 0.025mm thickness, as rolled
Expand
View Pricing
GF96730511
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 50 x 50mm, 0.25mm thickness, annealed
Expand
View Pricing
GF92612268
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 100 x 100mm, 0.25mm thickness, annealed
Expand
View Pricing
GF84316563
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 50mm disks, 0.25mm thickness, annealed
Expand
View Pricing
GF39247465
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 150 x 150mm, 0.25mm thickness, annealed
Expand
View Pricing
GF84269773
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 25 x 25mm, 0.25mm thickness, annealed
Expand
View Pricing
GF07069552
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 10mm disks, 0.025mm thickness, as rolled
Expand
View Pricing
GF56513055
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 15mm disks, 0.25mm thickness, annealed
Expand
View Pricing
GF35607977
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 25mm disks, 0.25mm thickness, annealed
Expand
View Pricing
GF51420406
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 4mm disks, 0.25mm thickness, annealed
Expand
View Pricing
GF03620887
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 1m coil, 0.25mm thickness, annealed
Expand
View Pricing
GF16996045
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 50mm disks, 0.025mm thickness, as rolled
Expand
View Pricing
GF37682208
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 100 x 100mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF94701450
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 50 x 50mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF72815687
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 0.5m coil, 0.25mm thickness, annealed
Expand
View Pricing
GF61735967
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 25mm disks, 0.025mm thickness, as rolled
Expand
View Pricing
GF10275869
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 150 x 150mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF88757612
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 25 x 25mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF47862579
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 50 x 50mm, 1mm thickness, annealed
Expand
View Pricing
GF33224431
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe,
foil, 6mm disks, 0.25mm thickness, annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2