Vanillic acid
Synonym(s):
4-Hydroxy-3-methoxybenzoic acid, Vanillic acid
Linear Formula:
HOC6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
168.15
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing