Valnemulin for peak identification
Synonym(s):
Valnemulin hydrochloride
Empirical Formula (Hill Notation):
C31H52N2O5S · HCl
CAS Number:
Molecular Weight:
601.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing