![Tris-O-(2-hydroxyethyl)rutin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/421/293/f3a0f7fc-8587-4f17-9ebc-59644022bcfd/640/f3a0f7fc-8587-4f17-9ebc-59644022bcfd.png)
Tris-O-(2-hydroxyethyl)rutin
![Tris-O-(2-hydroxyethyl)rutin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/421/293/f3a0f7fc-8587-4f17-9ebc-59644022bcfd/640/f3a0f7fc-8587-4f17-9ebc-59644022bcfd.png)
Synonym(s):
Trihydroxyethylrutin, 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Troxerutin
Empirical Formula (Hill Notation):
C33H42O19
CAS Number:
Molecular Weight:
742.68
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing