Skip to Content
Merck
Tris, Hydrochloride, Molecular Biology Grade Useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.

Tris, Hydrochloride, Molecular Biology Grade

Tris, Hydrochloride, Molecular Biology Grade Useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.
Synonym(s):
Trizma® hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride
Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60

Products

Product Category
Brand
Color
Formula Weight
pH Value
Physical Form
pK Value
Quality Segment