Skip to Content
Merck
Triflusulfuron-methyl certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Triflusulfuron-methyl

Triflusulfuron-methyl certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Methyl 2-[4-(dimethylamino)-6-(2,2,2-trifluoroethoxy)-1,3,5-triazin-2-ylaminocarbonylaminosulfonyl]-3-methylbenzoate
Empirical Formula (Hill Notation):
C17H19F3N6O6S
CAS Number:
Molecular Weight:
492.43

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade