Skip to Content
Merck
trans-2-Decenol natural (US), ≥97%, FG

trans-2-Decenol

trans-2-Decenol natural (US), ≥97%, FG
Synonym(s):
trans-2-Decen-1-ol
Linear Formula:
CH3(CH2)6CH=CHCH2OH
CAS Number:
Molecular Weight:
156.27

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing