Skip to Content
Merck
trans-2-Decenal analytical standard

trans-2-Decenal

trans-2-Decenal analytical standard
Linear Formula:
CH3(CH2)6CH=CHCHO
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing