Skip to Content
Merck
Tin(II) trifluoromethanesulfonate 97%

Tin(II) trifluoromethanesulfonate

Tin(II) trifluoromethanesulfonate 97%
Synonym(s):
Sn(OTf)2, Stannous trifluoromethanesulfonate, Tin(II) triflate
Linear Formula:
(CF3SO3)2Sn
CAS Number:
Molecular Weight:
416.85

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing