![Thioxanthone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/699/891/11b283e2-c756-4831-9042-ee4a605ca644/640/11b283e2-c756-4831-9042-ee4a605ca644.png)
Thioxanthone
![Thioxanthone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/699/891/11b283e2-c756-4831-9042-ee4a605ca644/640/11b283e2-c756-4831-9042-ee4a605ca644.png)
Synonym(s):
9-Oxothioxanthene, Thioxanthen-9-one
Empirical Formula (Hill Notation):
C13H8OS
CAS Number:
Molecular Weight:
212.27
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing