Thiopental
Synonym(s):
(±)-Thiopental, 5-Ethyl-5-(1-methylbutyl)-2-thiobarbituric acid, Pentothal
Empirical Formula (Hill Notation):
C11H18N2O2S
CAS Number:
Molecular Weight:
242.34
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing