Skip to Content
Merck
Thapsigargin InSolution, ≥97%, tumor-promoting sesquiterpene lactone

Thapsigargin

Thapsigargin InSolution, ≥97%, tumor-promoting sesquiterpene lactone
Synonym(s):
InSolution Thapsigargin
Empirical Formula (Hill Notation):
C34H50O12
CAS Number:
Molecular Weight:
650.75

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing