
Tetrakis(triphenylphosphine)palladium, polymer-bound

Synonym(s):
Palladium-tetrakis(triphenylphosphine), polymer-bound
Products
Brand
Physical Form
Markush Class
Markush Group
Chain Type
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing