![Tetrahydrocurcumin analytical standard](/deepweb/assets/sigmaaldrich/product/structures/264/766/12e8e13b-918f-4dd1-b87b-85ebc80a3f9f/640/12e8e13b-918f-4dd1-b87b-85ebc80a3f9f.png)
Tetrahydrocurcumin
![Tetrahydrocurcumin analytical standard](/deepweb/assets/sigmaaldrich/product/structures/264/766/12e8e13b-918f-4dd1-b87b-85ebc80a3f9f/640/12e8e13b-918f-4dd1-b87b-85ebc80a3f9f.png)
Synonym(s):
Tetrahydrocurcumin, 1,7-Bis(4-hydroxy-3-methoxyphenyl)-3,5-heptanedione
Empirical Formula (Hill Notation):
C21H24O6
CAS Number:
Molecular Weight:
372.41
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing