Skip to Content
Merck
Tetracontane analytical standard

Tetracontane

Tetracontane analytical standard
Synonym(s):
n-Tetracontane
Linear Formula:
CH3(CH2)38CH3
CAS Number:
Molecular Weight:
563.08
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing