Skip to Content
Merck
T0070907 ≥98% (HPLC)

T0070907

T0070907 ≥98% (HPLC)
Synonym(s):
2-Chloro-5-nitro-N-4-pyridinyl-benzamide, Benzamide, 2-chloro-5-nitro-N-4-pyridinyl-, T0070907
Empirical Formula (Hill Notation):
C12H8ClN3O3
CAS Number:
Molecular Weight:
277.66

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment