![Sulbactam Related Compound F / EP Impurity F certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/216/345/e6eea94a-4458-4f5d-9f96-a4e90c66eace/640/e6eea94a-4458-4f5d-9f96-a4e90c66eace.jpg)
Sulbactam Related Compound F / EP Impurity F
![Sulbactam Related Compound F / EP Impurity F certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/216/345/e6eea94a-4458-4f5d-9f96-a4e90c66eace/640/e6eea94a-4458-4f5d-9f96-a4e90c66eace.jpg)
Synonym(s):
(2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6,6-Dibromopenicillanic acid
Empirical Formula (Hill Notation):
C8H9Br2NO3S
CAS Number:
Molecular Weight:
359.03
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing