Skip to Content
Merck
Squalene ≥98%, liquid

Squalene

Squalene ≥98%, liquid
Synonym(s):
2,6,10,15,19,23-Hexamethyl-2,6,10,14,18,22-tetracosahexaene, Squalene
Linear Formula:
[(CH3)2C[=CHCH2CH2C(CH3)]2=CHCH2-]2
CAS Number:
Molecular Weight:
410.72

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique