Skip to Content
Merck
Sodium taurodeoxycholate hydrate BioXtra, ≥97% (TLC)

Sodium taurodeoxycholate hydrate

Sodium taurodeoxycholate hydrate BioXtra, ≥97% (TLC)
Synonym(s):
2-([3α,12α-Dihydroxy-24-oxo-5β-cholan-24-yl]amino)ethanesulfonic acid, Taurodeoxycholic acid sodium salt hydrate
Linear Formula:
C26H44NO6SNa · xH2O
CAS Number:
Molecular Weight:
521.69 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Product Line
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing