Skip to Content
Merck
Sodium caprylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sodium caprylate

Sodium caprylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Sodium octanoate, Caprylic acid sodium salt, Octanoic acid sodium salt, Sodium caprylate
Linear Formula:
CH3(CH2)6COONa
CAS Number:
Molecular Weight:
166.19

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Quality Segment
Application