Skip to Content
Merck
S-Methyl thioacetate natural, ≥96%, FG

S-Methyl thioacetate

S-Methyl thioacetate natural, ≥96%, FG
Empirical Formula (Hill Notation):
C3H6OS
CAS Number:
Molecular Weight:
90.14
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing