Skip to Content
Merck
Sisomicin Sulfate Pharmaceutical Secondary Standard; Certified Reference Material

Sisomicin sulfate

Sisomicin Sulfate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Sisomicin sulfate salt
Empirical Formula (Hill Notation):
C19H37N5O7 · 2.5H2O4S
CAS Number:
Molecular Weight:
692.72
EC Number:

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique