Skip to Content
Merck
Sinefungin 95% (HPLC), powder

Sinefungin

Sinefungin 95% (HPLC), powder
Synonym(s):
5′-Deoxy-5′-(1,4-diamino-4-carboxybutyl)adenosine, A-9145, Adenosylornithine, Antibiotic 32232RP, InSolution Sinefungin
Empirical Formula (Hill Notation):
C15H23N7O5
CAS Number:
Molecular Weight:
381.39

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-2 of 2