Skip to Content
Merck
Sibutramine hydrochloride monohydrate ≥98% (HPLC), solid

Sibutramine hydrochloride monohydrate

Sibutramine hydrochloride monohydrate ≥98% (HPLC), solid
Synonym(s):
1-(4-Chlorophenyl)-N,N-dimethyl-alpha-(2-methylpropyl)cyclobutanemethanamine hydrochloride monohydrate, Meridia, Reductil
Empirical Formula (Hill Notation):
C17H26ClN · HCl
CAS Number:
Molecular Weight:
316.31

Products

Shipping
Product Category
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing