Skip to Content
Merck
vibrant-m

Sevoflurane for system suitability

vibrant-m
Synonym(s):
Sevoflurane, 1,1,1,3,3,3-Hexafluoro-2-(fluoromethoxy)propane
Empirical Formula (Hill Notation):
C4H3F7O
CAS Number:
Molecular Weight:
200.05

Products

Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1