![sec-Butyllithium solution 1.4 M in cyclohexane](/deepweb/assets/sigmaaldrich/product/structures/762/170/16f919d7-07b4-4706-ac55-ae23ae9c43bb/640/16f919d7-07b4-4706-ac55-ae23ae9c43bb.png)
sec-Butyllithium solution
![sec-Butyllithium solution 1.4 M in cyclohexane](/deepweb/assets/sigmaaldrich/product/structures/762/170/16f919d7-07b4-4706-ac55-ae23ae9c43bb/640/16f919d7-07b4-4706-ac55-ae23ae9c43bb.png)
Synonym(s):
Lithium-2-butanide, s-BuLi
Linear Formula:
CH3CH2CH(CH3)Li
CAS Number:
Molecular Weight:
64.06
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing